Anti-CLEC12A Mouse Monoclonal Antibody [clone: 687317]

Supplier: R & D Systems
MAB2946
102885-526EA 639.2 USD
102885-526
Anti-CLEC12A Mouse Monoclonal Antibody [clone: 687317]
Antibodies
Type: Primary
Antigen: CLEC12A
Clonality: Monoclonal
Clone: 687317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR