Anti-ZBTB16 Mouse Monoclonal Antibody [clone: 6318100]

Supplier: R & D Systems
MAB2944
102885-524EA 639.2 USD
102885-524
Anti-ZBTB16 Mouse Monoclonal Antibody [clone: 6318100]
Antibodies
Type: Primary
Antigen: ZBTB16
Clonality: Monoclonal
Clone: 6318100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR