Anti-STX2 Rat Monoclonal Antibody [clone: 338824]

Supplier: R & D Systems
MAB2936
220020-191EA 577.02 USD
220020-191
Anti-STX2 Rat Monoclonal Antibody [clone: 338824]
Antibodies
Type: Primary
Antigen: STX2
Clonality: Monoclonal
Clone: 338824
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR