Anti-TGFBI Mouse Monoclonal Antibody [clone: 348506]

Supplier: R & D Systems
MAB2935
220020-190EA 763.57 USD
220020-190
Anti-TGFBI Mouse Monoclonal Antibody [clone: 348506]
Antibodies
Type: Primary
Antigen: TGFBI
Clonality: Monoclonal
Clone: 348506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR