Anti-VEGFA Mouse Monoclonal Antibody [clone: 23410]

Supplier: R & D Systems
MAB2931
220020-189EA 763.57 USD
220020-189
Anti-VEGFA Mouse Monoclonal Antibody [clone: 23410]
Antibodies
Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: 23410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR