Anti-MYD88 Rat Monoclonal Antibody [clone: 316628]

Supplier: R & D Systems
MAB29281
220020-187EA 594.37 USD
220020-187
Anti-MYD88 Rat Monoclonal Antibody [clone: 316628]
Antibodies
Type: Primary
Antigen: MYD88
Clonality: Monoclonal
Clone: 316628
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR