Anti-MYD88 Rat Monoclonal Antibody [clone: 316603]

Supplier: R & D Systems
MAB2928
220020-186EA 577.02 USD
220020-186
Anti-MYD88 Rat Monoclonal Antibody [clone: 316603]
Antibodies
Type: Primary
Antigen: MYD88
Clonality: Monoclonal
Clone: 316603
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR