Anti-CALR3 Mouse Monoclonal Antibody [clone: 321007]

Supplier: R & D Systems
MAB2927
102885-522EA 577.02 USD
102885-522
Anti-CALR3 Mouse Monoclonal Antibody [clone: 321007]
Antibodies
Type: Primary
Antigen: CALR3
Clonality: Monoclonal
Clone: 321007
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR