Anti-BMP15 Rat Monoclonal Antibody [clone: 317809]

Supplier: R & D Systems
MAB2925
220020-184EA 577.02 USD
220020-184
Anti-BMP15 Rat Monoclonal Antibody [clone: 317809]
Antibodies
Type: Primary
Antigen: BMP15
Clonality: Monoclonal
Clone: 317809
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR