Anti-IL18 Mouse Monoclonal Antibody [clone: 314820]

Supplier: R & D Systems
MAB2924
220020-182EA 561.11 USD
220020-182
Anti-IL18 Mouse Monoclonal Antibody [clone: 314820]
Antibodies
Type: Primary
Antigen: IL18
Clonality: Monoclonal
Clone: 314820
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR