Anti-RETNLB Mouse Monoclonal Antibody [clone: 250911]

Supplier: R & D Systems
MAB2917
220020-178EA 577.02 USD
220020-178
Anti-RETNLB Mouse Monoclonal Antibody [clone: 250911]
Antibodies
Type: Primary
Antigen: RETNLB
Clonality: Monoclonal
Clone: 250911
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR