Anti-IL19 Rat Monoclonal Antibody [clone: 350114]

Supplier: R & D Systems
MAB2915
220020-175EA 594.37 USD
220020-175
Anti-IL19 Rat Monoclonal Antibody [clone: 350114]
Antibodies
Type: Primary
Antigen: IL19
Clonality: Monoclonal
Clone: 350114
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR