Anti-ALPL Rat Monoclonal Antibody [clone: 388828]

Supplier: R & D Systems
MAB29091
102885-520EA 594.37 USD
102885-520
Anti-ALPL Rat Monoclonal Antibody [clone: 388828]
Antibodies
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: 388828
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR