Anti-ALPL Rat Monoclonal Antibody [clone: 388816]

Supplier: R & D Systems
MAB2909
220020-173EA 594.37 USD
220020-173
Anti-ALPL Rat Monoclonal Antibody [clone: 388816]
Antibodies
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: 388816
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR