Anti-wnvNS3 Protease NS2b Cofactor Mouse Monoclonal Antibody [clone: 345712]

Supplier: R & D Systems
MAB29071
220020-172EA 561.11 USD
220020-172
Anti-wnvNS3 Protease NS2b Cofactor Mouse Monoclonal Antibody [clone: 345712]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: wnvNS3 Protease NS2b Cofactor
Antigen Synonyms:
Clonality: Monoclonal
Clone: 345712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR