Anti-wnvNS3 Protease Mouse Monoclonal Antibody [clone: 345713]

Supplier: R & D Systems
MAB2907
220020-171EA 566.89 USD
220020-171
Anti-wnvNS3 Protease Mouse Monoclonal Antibody [clone: 345713]
Antibodies
Type: Primary
Antigen: wnvNS3 Protease
Clonality: Monoclonal
Clone: 345713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR