Anti-GZMB Mouse Monoclonal Antibody [clone: 351927]

Supplier: R & D Systems
MAB2906
220020-170EA 594.37 USD
220020-170
Anti-GZMB Mouse Monoclonal Antibody [clone: 351927]
Antibodies
Type: Primary
Antigen Symbol: GZMB
Antigen Name: granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1)
Antigen Synonyms: CCPI, CGL-1, CGL1, CSP-B, CSPB, CTLA1, CTSGL1, HLP, SECT
Clonality: Monoclonal
Clone: 351927
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR