Anti-GZMA Mouse Monoclonal Antibody [clone: 356412]

Supplier: R & D Systems
MAB29051
220020-169EA 633.42 USD
220020-169
Anti-GZMA Mouse Monoclonal Antibody [clone: 356412]
Antibodies
Type: Primary
Antigen Symbol: GZMA
Antigen Name: granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3)
Antigen Synonyms: CTLA3, HFSP
Clonality: Monoclonal
Clone: 356412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR