Anti-IL9R Mouse Monoclonal Antibody [clone: 33449]

Supplier: R & D Systems
MAB2901
220020-167EA 772.25 USD
220020-167
Anti-IL9R Mouse Monoclonal Antibody [clone: 33449]
Antibodies
Type: Primary
Antigen: IL9R
Clonality: Monoclonal
Clone: 33449
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR