Anti-ACY1 Rat Monoclonal Antibody [clone: 475626]

Supplier: R & D Systems
MAB2900
220020-166EA 577.02 USD
220020-166
Anti-ACY1 Rat Monoclonal Antibody [clone: 475626]
Antibodies
Type: Primary
Antigen: ACY1
Clonality: Monoclonal
Clone: 475626
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR