Anti-CPB1 Mouse Monoclonal Antibody [clone: 438806]

Supplier: R & D Systems
MAB2897
220020-164EA 577.02 USD
220020-164
Anti-CPB1 Mouse Monoclonal Antibody [clone: 438806]
Antibodies
Type: Primary
Antigen: CPB1
Clonality: Monoclonal
Clone: 438806
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR