Anti-CPA2 Mouse Monoclonal Antibody [clone: 384023]

Supplier: R & D Systems
MAB28961
220020-163EA 561.11 USD
220020-163
Anti-CPA2 Mouse Monoclonal Antibody [clone: 384023]
Antibodies
Type: Primary
Antigen: CPA2
Clonality: Monoclonal
Clone: 384023
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR