Anti-CPA2 Mouse Monoclonal Antibody [clone: 384014]

Supplier: R & D Systems
MAB2896
220020-162EA 763.57 USD
220020-162
Anti-CPA2 Mouse Monoclonal Antibody [clone: 384014]
Antibodies
Type: Primary
Antigen: CPA2
Clonality: Monoclonal
Clone: 384014
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR