Anti-MEP1B Rat Monoclonal Antibody [clone: 289703]

Supplier: R & D Systems
MAB28952
220020-161EA 577.02 USD
220020-161
Anti-MEP1B Rat Monoclonal Antibody [clone: 289703]
Antibodies
Type: Primary
Antigen: MEP1B
Clonality: Monoclonal
Clone: 289703
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR