Anti-MEP1B Rat Monoclonal Antibody [clone: 289731]

Supplier: R & D Systems
MAB28951
102885-518EA 594.37 USD
102885-518
Anti-MEP1B Rat Monoclonal Antibody [clone: 289731]
Antibodies
Type: Primary
Antigen: MEP1B
Clonality: Monoclonal
Clone: 289731
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR