Anti-STAT2 Rabbit Monoclonal Antibody [clone: 1021D]

Supplier: R & D Systems
MAB2890
102885-516EA 608.83 USD
102885-516
Anti-STAT2 Rabbit Monoclonal Antibody [clone: 1021D]
Antibodies
Simple Western

Type: Primary
Antigen: STAT2
Clonality: Monoclonal
Clone: 1021D
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR