Anti-BCL10 Mouse Monoclonal Antibody [clone: 384202]

Supplier: R & D Systems
MAB2889
220020-159EA 561.11 USD
220020-159
Anti-BCL10 Mouse Monoclonal Antibody [clone: 384202]
Antibodies
Type: Primary
Antigen: BCL10
Clonality: Monoclonal
Clone: 384202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR