Anti-EPAS1 Mouse Monoclonal Antibody [clone: 318404]

Supplier: R & D Systems
MAB2886
220020-157EA 594.37 USD
220020-157
Anti-EPAS1 Mouse Monoclonal Antibody [clone: 318404]
Antibodies
Type: Primary
Antigen: EPAS1
Clonality: Monoclonal
Clone: 318404
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR