Anti-OMD Mouse Monoclonal Antibody [clone: 806001]

Supplier: R & D Systems
MAB2884
102885-514EA 577.02 USD
102885-514
Anti-OMD Mouse Monoclonal Antibody [clone: 806001]
Antibodies
Type: Primary
Antigen: OMD
Clonality: Monoclonal
Clone: 806001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR