Anti-KIR3DL2 Mouse Monoclonal Antibody [clone: 539304]

Supplier: R & D Systems
MAB2878
102885-510EA 600.15 USD
102885-510
Anti-KIR3DL2 Mouse Monoclonal Antibody [clone: 539304]
Antibodies
Type: Primary
Antigen: KIR3DL2
Clonality: Monoclonal
Clone: 539304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR