Anti-FLRT2 Mouse Monoclonal Antibody [clone: 367205]

Supplier: R & D Systems
MAB2877
220020-154EA 577.02 USD
220020-154
Anti-FLRT2 Mouse Monoclonal Antibody [clone: 367205]
Antibodies
Type: Primary
Antigen Symbol: FLRT2
Antigen Name: fibronectin leucine rich transmembrane protein 2
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 367205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR