Anti-EPYC Rat Monoclonal Antibody [clone: 340908]

Supplier: R & D Systems
MAB2875
220020-153EA 561.11 USD
220020-153
Anti-EPYC Rat Monoclonal Antibody [clone: 340908]
Antibodies
Type: Primary
Antigen: EPYC
Clonality: Monoclonal
Clone: 340908
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR