Anti-FAM3D Mouse Monoclonal Antibody [clone: 358203]

Supplier: R & D Systems
MAB2869
220020-148EA 594.37 USD
220020-148
Anti-FAM3D Mouse Monoclonal Antibody [clone: 358203]
Antibodies
Type: Primary
Antigen Symbol: FAM3D
Antigen Name: family with sequence similarity 3, member D
Antigen Synonyms: EF7, OIT1
Clonality: Monoclonal
Clone: 358203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR