Anti-FAM3C Rat Monoclonal Antibody [clone: 372018]

Supplier: R & D Systems
MAB2868
220020-147EA 561.11 USD
220020-147
Anti-FAM3C Rat Monoclonal Antibody [clone: 372018]
Antibodies
Type: Primary
Antigen Symbol: FAM3C
Antigen Name: family with sequence similarity 3, member C
Antigen Synonyms: D6Wsu176e, Ilei
Clonality: Monoclonal
Clone: 372018
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR