Anti-FAM3B Mouse Monoclonal Antibody [clone: 365505]

Supplier: R & D Systems
MAB2866
220020-146EA 577.02 USD
220020-146
Anti-FAM3B Mouse Monoclonal Antibody [clone: 365505]
Antibodies
Type: Primary
Antigen Symbol: FAM3B
Antigen Name: family with sequence similarity 3, member B
Antigen Synonyms: 2-21, C21orf11, C21orf76, ORF9, PANDER, PRED44
Clonality: Monoclonal
Clone: 365505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR