Anti-FAM3A Mouse Monoclonal Antibody [clone: 358112]

Supplier: R & D Systems
MAB2865
220020-145EA 561.11 USD
220020-145
Anti-FAM3A Mouse Monoclonal Antibody [clone: 358112]
Antibodies
Type: Primary
Antigen Symbol: FAM3A
Antigen Name: family with sequence similarity 3, member A
Antigen Synonyms: 2.19, DLD, DXS560S, XAP-7
Clonality: Monoclonal
Clone: 358112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR