Anti-SOX10 Mouse Monoclonal Antibody [clone: 20B7]

Supplier: R & D Systems
MAB2864
220020-144EA 655.11 USD
220020-144
Anti-SOX10 Mouse Monoclonal Antibody [clone: 20B7]
Antibodies
Type: Primary
Antigen: SOX10
Clonality: Monoclonal
Clone: 20B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR