Anti-SLITRK4 Mouse Monoclonal Antibody [clone: 319514]

Supplier: R & D Systems
MAB2860
220020-142EA 577.02 USD
220020-142
Anti-SLITRK4 Mouse Monoclonal Antibody [clone: 319514]
Antibodies
Type: Primary
Antigen: SLITRK4
Clonality: Monoclonal
Clone: 319514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR