Anti-SIGLEC6 Mouse Monoclonal Antibody [clone: 767329]

Supplier: R & D Systems
MAB2859
102885-508EA 655.11 USD
102885-508
Anti-SIGLEC6 Mouse Monoclonal Antibody [clone: 767329]
Antibodies
Type: Primary
Antigen: SIGLEC6
Clonality: Monoclonal
Clone: 767329
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR