Anti-PRSS34 Rat Monoclonal Antibody [clone: 451421]

Supplier: R & D Systems
MAB2857
220020-140EA 577.02 USD
220020-140
Anti-PRSS34 Rat Monoclonal Antibody [clone: 451421]
Antibodies
Type: Primary
Antigen: PRSS34
Clonality: Monoclonal
Clone: 451421
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR