Anti-MAP2K2 Mouse Monoclonal Antibody [clone: 300317]

Supplier: R & D Systems
MAB2855
220020-138EA 577.02 USD
220020-138
Anti-MAP2K2 Mouse Monoclonal Antibody [clone: 300317]
Antibodies
Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 300317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR