Anti-LUM Mouse Monoclonal Antibody [clone: 669401]

Supplier: R & D Systems
MAB28461
102885-504EA 594.37 USD
102885-504
Anti-LUM Mouse Monoclonal Antibody [clone: 669401]
Antibodies
Type: Primary
Antigen: LUM
Clonality: Monoclonal
Clone: 669401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR