Anti-LUM Mouse Monoclonal Antibody [clone: 358022]

Supplier: R & D Systems
MAB2846
220020-134EA 594.37 USD
220020-134
Anti-LUM Mouse Monoclonal Antibody [clone: 358022]
Antibodies
Type: Primary
Antigen: LUM
Clonality: Monoclonal
Clone: 358022
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR