Anti-Oxidized PTP Active Site Mouse Monoclonal Antibody [clone: 335636]

Supplier: R & D Systems
MAB2844
220020-132EA 655.11 USD
220020-132
Anti-Oxidized PTP Active Site Mouse Monoclonal Antibody [clone: 335636]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Oxidized PTP Active Site
Antigen Synonyms:
Clonality: Monoclonal
Clone: 335636
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR