Anti-IL2RG Mouse Monoclonal Antibody [clone: 633162]

Supplier: R & D Systems
MAB2842
102885-502EA 623.29 USD
102885-502
Anti-IL2RG Mouse Monoclonal Antibody [clone: 633162]
Antibodies
Type: Primary
Antigen: IL2RG
Clonality: Monoclonal
Clone: 633162
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR