Anti-IL2RG Mouse Monoclonal Antibody [clone: 31134]

Supplier: R & D Systems
MAB2841
220020-131EA 772.25 USD
220020-131
Anti-IL2RG Mouse Monoclonal Antibody [clone: 31134]
Antibodies
Type: Primary
Antigen: IL2RG
Clonality: Monoclonal
Clone: 31134
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR