Anti-PPP3CA Rat Monoclonal Antibody [clone: 302202]

Supplier: R & D Systems
MAB2839
220020-129EA 594.37 USD
220020-129
Anti-PPP3CA Rat Monoclonal Antibody [clone: 302202]
Antibodies
Type: Primary
Antigen: PPP3CA
Clonality: Monoclonal
Clone: 302202
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR