Anti-SPARCL1 Rat Monoclonal Antibody [clone: 348905]

Supplier: R & D Systems
MAB2836
220020-127EA 594.37 USD
220020-127
Anti-SPARCL1 Rat Monoclonal Antibody [clone: 348905]
Antibodies
Type: Primary
Antigen: SPARCL1
Clonality: Monoclonal
Clone: 348905
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR