Anti-STC2 Rat Monoclonal Antibody [clone: 348217]

Supplier: R & D Systems
MAB2830
220020-124EA 561.11 USD
220020-124
Anti-STC2 Rat Monoclonal Antibody [clone: 348217]
Antibodies
Type: Primary
Antigen: STC2
Clonality: Monoclonal
Clone: 348217
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR