Anti-SLAMF9 Rat Monoclonal Antibody [clone: 330003]

Supplier: R & D Systems
MAB2829
220020-122EA 594.37 USD
220020-122
Anti-SLAMF9 Rat Monoclonal Antibody [clone: 330003]
Antibodies
Type: Primary
Antigen: SLAMF9
Clonality: Monoclonal
Clone: 330003
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR